missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tungsten foil, 1.0mm (0.04in) thick, 99.95% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 013981FY

 View more versions of this product

Catalog No. AA13981FY



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tungsten
Silver Gray foil
99.95% (Metals basis)
MFCD00011461
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
[W]
183.84
CHEBI:27998
7440-33-7
19.3g/cm3
W
50 x 100 mm
WFKWXMTUELFFGS-UHFFFAOYSA-N
tungsten
23964
Foil

Chemical Identifiers

7440-33-7
183.84
WFKWXMTUELFFGS-UHFFFAOYSA-N
23964
tungsten
W
MFCD00011461
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
CHEBI:27998
[W]
Safety and Handling

Safety and Handling

EINECSNumber : 231-143-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only