missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tungsten Foil, 0.5mm (0.02 in.) thick, 99.95% (Metals basis), Alfa Aesar™

Form: Approx. 12g/25 x 50mm

Manufacturer:  Thermo Fisher Scientific 10414HB

 View more versions of this product

Catalog No. AA10414HB



Specifications

Specifications

Tungsten
7440-33-7
99.95% (Metals basis)
MFCD00011461
WFKWXMTUELFFGS-UHFFFAOYSA-N
tungsten
23964
Foil, 0.5mm (0.02 in.) thick
300 x 300mm
Silver Gray foil
W
4+ ion, ion, powder, tungsten, elemental, tungsten, insoluble compounds, tungsten, soluble compounds, unii-v9306cxo6g, va, wolfram, wolframium
[W]
183.84
CHEBI:27998
Silver to Gray

Chemical Identifiers

7440-33-7
183.84
WFKWXMTUELFFGS-UHFFFAOYSA-N
23964
tungsten
W
MFCD00011461
4+ ion, ion, powder, tungsten, elemental, tungsten, insoluble compounds, tungsten, soluble compounds, unii-v9306cxo6g, va, wolfram, wolframium
CHEBI:27998
[W]
Safety and Handling

Safety and Handling

EINECSNumber : 231-143-9

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents