missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tungsten Foil, 0.1mm (0.004 in.) thick, 99.95% (Metals basis), Alfa Aesar™

Form: Approx. 4.8g/50 x 50mm

Manufacturer:  Alfa Aesar 10416HW

 View more versions of this product

Catalog No. AA10416HW



Specifications

Specifications

Tungsten
7440-33-7
99.95% (Metals basis)
MFCD00011461
WFKWXMTUELFFGS-UHFFFAOYSA-N
tungsten
23964
Foil, 0.1mm (0.004 in.) thick
150 x 300mm
Silver Gray foil
W
4+ ion
[W]
183.84
CHEBI:27998
Silver to Gray

Chemical Identifiers

7440-33-7
183.84
WFKWXMTUELFFGS-UHFFFAOYSA-N
23964
tungsten
W
MFCD00011461
4+ ion
CHEBI:27998
[W]
Documents