missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tungsten carbide Cobalt, 99% (metals basis)

Manufacturer:  Alfa Aesar™ 2295530

 View more versions of this product

Catalog No. AA2295530



Specifications

Specifications

WC:Co; 94:6 wt%
250g
99%
CHCoW
UN3178
IKEHVRMRUVYPGG-UHFFFAOYSA-N
C#[W].[Co]
255.792
Powder
Tungsten carbide Cobalt
12774-15-1
(metals basis)
MFCD00064847
−325 Mesh
Odorless
cobalt;methylidynetungsten
56846058
Gray

Chemical Identifiers

12774-15-1
255.792
IKEHVRMRUVYPGG-UHFFFAOYSA-N
cobalt;methylidynetungsten
CHCoW
MFCD00064847
56846058
C#[W].[Co]
Safety and Handling

Safety and Handling

GHS H Statement
H334-H228-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable solid.
May cause an allergic skin reaction.

GHS P Statement
P210-P284a-P261-P280-P342+P311-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.

TSCA : TSCA

SDS Documents