Please login to your online account to display your discounted pricing

Alfa Aesar™ Tungsten carbide Cobalt, 99% (metals basis)

Chemical Identifiers

Chemical Name or Material Tungsten carbide Cobalt
CAS 12774-15-1
MDL Number MFCD00064847
UN Number UN3178
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA22955A1

  SDS
alfa aesar™
22955A1
1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA2295530

  SDS
alfa aesar™
2295530
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay Percent Notes (metals basis)
Health Hazard 2 GHS H Statement
H334-H228-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable solid.
May cause an allergic skin reaction.
Quantity 1kg
Health Hazard 1 Danger
MDL Number MFCD00064847
CAS 12774-15-1
Health Hazard 3 GHS P Statement
P210-P284a-P261-P280-P342+P311-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Odor Odorless
Assay 99%
UN Number UN3178
Chemical Name or Material Tungsten carbide Cobalt
Physical Form Powder
TSCA TSCA
Avg. Mol. Wt. or Mol. Wt. Range WC:Co; 94:6 wt%
Mesh Size −325 Mesh
Color Dark gray
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P284a-P261-P280-P342+P311-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H228-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable solid.
May cause an allergic skin reaction.