Please login to your online account to display your discounted pricing

Tungsten Boride, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Tungsten Boride
CAS 12007-10-2
Physical Form -325 Mesh Powder
MDL Number MFCD00049708
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA8816614

 
alfa aesar
8816614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form -325 Mesh Powder
Quantity 25g
Chemical Name or Material Tungsten Boride
MDL Number MFCD00049708
CAS 12007-10-2