Please login to your online account to display your discounted pricing

TUG 424, Tocris™

FFA1 (GPR40) agonist

$905.98 - $905.98

Chemical Identifiers

Chemical Name or Material TUG 424
Assay Percent Range >99%
Molecular Formula C18H16O2
Formula Weight 264.32
Synonym 4-[2-(2-methylphenyl)ethynyl]-benzenepropanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

379550

 
r & d systems
RDS379550
50mg Each for $905.98
Description & Specifications

Specifications

Formula Weight 264.32
Quantity 50mg
Chemical Name or Material TUG 424
Assay Percent Range >99%
Molecular Formula C18H16O2
CAS 1082058-99-8
Synonym 4-[2-(2-methylphenyl)ethynyl]-benzenepropanoic acid
Recommended Storage Store at +4°C