Bio-Serv™ Tuff Turf Foraging Boards
Manufacturer:  Bio-Serv™ F9112
 View more versions of this product
Catalog No. 14727036
Description
Description
  • Made of Stainless Steel
  • Available in 2 sizes
  • Extra-durable artificial turf is securely anchored
    by stainless steel bars and lock nuts
  • Texture of turf hides small treats increasing foraging time
  • Tray has drainage holes for easy cleaning
  • Replacement turf pads available
  • Autoclavable (Board Only)
  • Cagewasher safe
Specifications
Specifications
6 x 6in.
Stainless Steel
Large
Foraging Board
Documents