Please login to your online account to display your discounted pricing

Tubulin Mouse, Clone: 2G10-TB3, eBioscience™

Mouse Monoclonal Antibody

$131.95 - $294.35

Specifications

Antigen Tubulin
Clone 2G10-TB3
Host Species Murine
Gene Alias TUBB3
Species Reactivity Bovine, Human, Mouse, Rat
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122676

 
life technologies
LS14451080
25μg Each for $131.95

501122677

 
life technologies
LS14451082
100μg Each for $294.35
Description & Specifications

Specifications

Antigen Tubulin
Clone 2G10-TB3
Host Species Murine
Gene Alias TUBB3
Species Reactivity Bovine, Human, Mouse, Rat
Applications Immunocytochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Microscopy
Applications Western Blot
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The 2G10 monoclonal antibody recognizes beta tubulin class III, a member of the large family of tubulins that are components of the microtubules of the cytoskeletal network. Beta tubulin class III expression is exclusively found in neuronal cells of adult mammals. Although expression has been mainly found in neuronal cells, during fetal development, transient expression has been documented in both glial and neuronal precursors localized to the subventricular zone. Identification of beta tubulin class III expression is useful as a structural marker for developing and mature neural cells. Given the high degree of conservation of this protein across species, the 2G10 clone is expected to recognize human, mouse, rat, and bovine beta tubulin class III.