Please login to your online account to display your discounted pricing

tubulin Mouse, Biotin, Clone: DM1A, eBioscience™

Mouse Monoclonal Antibody

$131.95 - $323.35

Specifications

Antigen tubulin
Clone DM1A
Host Species Murine
Gene Alias alpha-Tubulin, a-Tubulin
Species Reactivity Canine, Human, Monkey, Mouse, Pig, Rat
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011861

 
life technologies
LS13450280
25μg Each for $131.95

5011862

 
life technologies
LS13450282
100μg Each for $323.35
Description & Specifications

Specifications

Antigen tubulin
Clone DM1A
Host Species Murine
Gene Alias alpha-Tubulin, a-Tubulin
Species Reactivity Canine, Human, Monkey, Mouse, Pig, Rat
Applications Immunocytochemistry
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The monoclonal antibody DM1a recognizes the 50kDa cytoskeletal protein α tubulin in a variety of species (human, mouse, rat, monkey, dog, pig, bovine, goat, hamster, guinea pig, kangaroo, amphibians, sea urchin, yeast and tobacco plants). Tubulin, the major component of microtubules, is a dimeric protein consisting of an alpha and beta subunit. Tubulin is a GTP-binding protein that can be modified by phosphorylation and acetylation resulting in assembly (polymerization) or disassembly (depolymerization). The dynamic nature of microtubules is most evident in the mitotic apparatus. The DM1a antibody recognizes the C-terminal end of the α tubulin isoform (amino acids 426-430).