Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Tubulin-Β Ab-6, Mouse Monoclonal Antibody


Provide accurate, reproducible results with the Thermo Scientific™ Tubulin-Β Ab-6, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1226PABX

 View more versions of this product

Catalog No. MS1226PABX

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Bovine
Cross Reactivity Chicken
Cross Reactivity Frog
Cross Reactivity Human
Cross Reactivity Moth
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Wheat
Isotype IgG3
Format Concentrated
Immunogen Purified tubulin
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Regulatory Status RUO
Clone TBN06; same as Tub 2.5
Quantity 200µL
Host Species Mouse
Antigen Tubulin-beta Ab-6
Type Primary
Formulation Ab without BSA/Azide (Frozen)
Conjugate Unlabeled
Research Discipline Cytoskeletal Protein
Concentration 1mg/mL
Classification Monoclonal
Item Description 200uL; 1mg/mL; Purified w/o BSA/ azide

Provide accurate, reproducible results with the Thermo Scientific™ Tubulin-Β Ab-6, Mouse Monoclonal Antibody.

Microtubules, the major cytoskeletal elements found in all eukaryotic cells, consist of Tubulin, which is a dimer of two 55kDa subunits: alpha and Beta. Microtubules play key roles in chromosome segregation in mitosis, intracellular transport, ciliary and flagellar bending, and structural support of the cytoskeleton.

Host Species: Mouse

Clone: TBN06; same as Tub 2.5

Isotype: IgG3

Species Reactivity: Human, Cow, Rabbit, Mouse, Chicken, Rat, Frog, Moth and Wheat. Others not known.

Epitope: Not determined

Immunogen: Purified tubulin

Molecular Weight: 55kDa

Positive Control: Ls174T, MAD109 cells; Skin or lung

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO