Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Tubulin-Β Ab-6, Mouse Monoclonal Antibody


Provide accurate, reproducible results with the Thermo Scientific™ Tubulin-Β Ab-6, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1226P1ABX

 View more versions of this product

Catalog No. MS1226P1ABX

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Host Species Mouse
Cross Reactivity Bovine
Cross Reactivity Chicken
Cross Reactivity Frog
Cross Reactivity Human
Cross Reactivity Moth
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Wheat
Antigen Tubulin-beta Ab-6
Quantity 100µL
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Isotype IgG3
Immunogen Purified tubulin
Format Concentrated
Clone TBN06; same as Tub 2.5
Conjugate Unlabeled
Formulation Ab without BSA/Azide (Frozen)
Classification Monoclonal
Research Discipline Cytoskeletal Protein
Type Primary
Concentration 1mg/mL

Provide accurate, reproducible results with the Thermo Scientific™ Tubulin-Β Ab-6, Mouse Monoclonal Antibody.

Microtubules, the major cytoskeletal elements found in all eukaryotic cells, consist of Tubulin, which is a dimer of two 55kDa subunits: alpha and Beta. Microtubules play key roles in chromosome segregation in mitosis, intracellular transport, ciliary and flagellar bending, and structural support of the cytoskeleton.

Host Species: Mouse

Clone: TBN06; same as Tub 2.5

Isotype: IgG3

Species Reactivity: Human, Cow, Rabbit, Mouse, Chicken, Rat, Frog, Moth and Wheat. Others not known.

Epitope: Not determined

Immunogen: Purified tubulin

Molecular Weight: 55kDa

Positive Control: Ls174T, MAD109 cells; Skin or lung

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO