missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tubocurarine Chloride Pentahydrate, TCI America™

$65.70 - $415.56

Chemical Identifiers

CAS 6989-98-6
Molecular Formula C37H42Cl2N2O6
Molecular Weight (g/mol) 681.651
MDL Number MFCD00013471
InChI Key GXFZCDMWGMFGFL-KKXMJGKMSA-N
Synonym Curare
PubChem CID 16051918
SMILES CN1CCC2=CC(=C3C=C2C1CC4=CC=C(C=C4)OC5=C6C(CC7=CC(=C(C=C7)O)O3)[N+](CCC6=CC(=C5O)OC)(C)C)OC.Cl.[Cl-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C0433100MG View Documents TCI America
C0433100MG
100mg Each for $65.70
 
C04331G View Documents TCI America
C04331G
1g Each for $415.56
 
Specifications

Chemical Identifiers

6989-98-6
681.651
GXFZCDMWGMFGFL-KKXMJGKMSA-N
16051918
C37H42Cl2N2O6
MFCD00013471
Curare
CN1CCC2=CC(=C3C=C2C1CC4=CC=C(C=C4)OC5=C6C(CC7=CC(=C(C=C7)O)O3)[N+](CCC6=CC(=C5O)OC)(C)C)OC.Cl.[Cl-]

Specifications

Undesignated
100mg
C37H42Cl2N2O6
1544
GXFZCDMWGMFGFL-KKXMJGKMSA-N
681.651
681.65
≥98.0% (HPLC,T)
268°C
6989-98-6
MFCD00013471
Curare
CN1CCC2=CC(=C3C=C2C1CC4=CC=C(C=C4)OC5=C6C(CC7=CC(=C(C=C7)O)O3)[N+](CCC6=CC(=C5O)OC)(C)C)OC.Cl.[Cl-]
16051918
Crystal-Powder at 20°C
Tubocurarine Chloride Pentahydrate
Safety and Handling

Safety and Handling

RTECSNumber : YO5100000

TSCA : No