missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Tube Rocker Dimple Rubber Mat
Accessory for use with Digital Tube Rocker, 120V and Digital Tube Rocker, 230V
Supplier: Thermo Scientific™ 88882159
Description
Help ensure peace of mind when using this accessory with the Thermo Scientific™ Digital Tube Rocker.
Specifications
Tubes | |
32.3 x 13.3 cm (12.7 x 5 in.) | |
Tube Rocker Stacking Bars (88882158), Tube Rocker Stacking Bars (88882160), Variety of tubes | |
7 |
30 mm | |
Tube Rocker Dimple Rubber Mat | |
Digital Tube Rocker, 120V (88882017)Digital Tube Rocker, 230V (88882018) |