Labcon North America TUBE 50ML PP STERILE 500CS
Supplier: Labcon North America 31913350189
Tube, Centrifuge; Labcon; Polypropylene; With attached flat cap in a proprietary cardboard rack; Sterile; Printed graduations; Capacity: 50mL; 25/rack; 20 racks/case