Please login to your online account to display your discounted pricing

TTNPB, Tocris


Retinoic acid analog; RAR agonist

Quantity 10mg

Manufacturer: r & d systems  RDS076110

 View more versions of this product

Catalog No. 076110

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 348.48
Chemical Name or Material TTNPB
Assay Percent Range >98%
Molecular Formula C24H28O2
CAS 71441-28-6
Synonym 4-[(E)-2-(5,6,7,8-Tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl)-1-propenyl]benzoic acid
Recommended Storage Desiccate at -20°C

Specifications

Chemical Name or Material TTNPB
Assay Percent Range >98%
Molecular Formula C24H28O2
Formula Weight 348.48
Synonym 4-[(E)-2-(5,6,7,8-Tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl)-1-propenyl]benzoic acid
Accessories

Desiccate at -20°C