Invitrogen™ TTK Recombinant Human Protein DFS Item
TTK is a serine/threonine kinase closely related to SPK1, PIM 1 and CDC2 serine/threonine kinases.
Manufacturer:  Invitrogen™ LSPV4071
 View more versions of this product
Catalog No. PV4071
Description
Description
TTK is a dual-specificity kinase, which can also phosphorylate tyrosine.
Specifications
Specifications
NP_003309
Frozen
126.3kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
TTK Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
TTK
Baculovirus
Human
GST Tag
Documents