Please login to your online account to display your discounted pricing

TSLP Receptor Mouse anti-Human, PerCP-eFluor 710, Clone: eBio1A6 (1A6), eBioscience™

Mouse Monoclonal Antibody

$216.05 - $506.05

Specifications

Antigen TSLP Receptor
Clone eBio1A6 (1A6)
Host Species Murine
Gene Alias CRLF-2, thymic stromal derived lymphopoietin receptor
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016065

 
life technologies
LS46549941
25 Tests Each for $216.05

5016066

 
life technologies
LS46549942
100 Tests Each for $506.05
Description & Specifications

Specifications

Antigen TSLP Receptor
Clone eBio1A6 (1A6)
Host Species Murine
Gene Alias CRLF-2, thymic stromal derived lymphopoietin receptor
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The eBio1A6 monoclonal antibody reacts with human thymic stromal-derived lymphopoietin receptor (TSLPR). TSLPR is an approximately 50kDa protein with significant similarity to the common gamma-chain. TSLPR complexes with IL-7R alpha (CD127) to form the high affinity receptor that binds thymic stromal-derived lymphopoietin (TSLP). Human TSLPR is expressed by monocytes and CD11c+ dendritic cells, and TSLP binding induces the expression of the Th2 cell-attracting chemokines CCL17 and CCL22. Furthermore, the TSLPR-induced activation of dendritic cells indirectly results in the increased secretion of Th cytokines IL-4, -5 and -13, which may be necessary for the regulation of CD4+ T cell homeostasis. In mice, deficiency of TSLPR has no effect on lymphocyte numbers, whereas double deficiency of TSLPR and common gamma-chain results in fewer lymphocytes than seen in mice deficient in the common gamma-chain alone. The eBio1A6 monoclonal antibody is able to cross-block binding of another anti-human TSLPR monoclonal antibody, 1D8.