Please login to your online account to display your discounted pricing

TSLP Receptor Mouse anti-Human, APC, Clone: 1A6, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $433.55

Specifications

Antigen TSLP Receptor
Clone eBio1A6 (1A6)
Host Species Murine
Gene Alias thymic stromal derived lymphopoietin receptor, CRLF-2
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123134

 
life technologies
LS17549941
25 Tests Each for $172.55

501129286

 
life technologies
LS17549942
100 Tests Each for $433.55
Description & Specifications

Specifications

Antigen TSLP Receptor
Clone eBio1A6 (1A6)
Host Species Murine
Gene Alias thymic stromal derived lymphopoietin receptor, CRLF-2
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Allophycocyanin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.06μg)/test

The eBio1A6 monoclonal antibody reacts with human thymic stromal-derived lymphopoietin receptor (TSLPR). TSLPR is an approximately 50kDa protein with significant similarity to the common gamma-chain. TSLPR complexes with IL-7R alpha (CD127) to form the high affinity receptor that binds thymic stromal-derived lymphopoietin (TSLP). Human TSLPR is expressed by monocytes and CD11c+ dendritic cells, and TSLP binding induces the expression of the Th2 cell-attracting chemokines CCL17 and CCL22. Furthermore, the TSLPR-induced activation of dendritic cells indirectly results in the increased secretion of Th cytokines IL-4, -5 and -13, which may be necessary for the regulation of CD4+ T cell homeostasis. In mice, deficiency of TSLPR has no effect on lymphocyte numbers, whereas double deficiency of TSLPR and common gamma-chain results in fewer lymphocytes than seen in mice deficient in the common gamma-chain alone. The eBio1A6 monoclonal antibody is able to cross-block binding of another anti-human TSLPR monoclonal antibody, 1D8.