missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ TSG Series Countertop Refrigerator

Innovative refrigeration solutions for healthcare and research needs

Manufacturer:  Thermo Scientific™ TSG205SA

Catalog No. TSG205SA



Description

Description

The Thermo Scientific™ TSG Series Countertop Refrigerator, powered by Phononic solid state refrigeration technology, has been designed to meet the rigorous and demanding needs of healthcare, research and industrial applications where  solid performance is required, in support of breastmilk and medication storage, analyzer kits and reagents, and even cell culture media.

Key features of the TSG205SA Countertop Refrigerator:

  • Solid state refrigeration technology
  • Whisper quiet operation leading to a more comfortable work environment supporting clinical and healthcare needs
  • Increased capacities due to fewer refrigeration components means you can store more in less space
  • Green refrigerants supporting global initiatives to reduce green house gas emissions
  • Full alarm set, including hi/lo temperature, door ajar and power fail to support requirements for medical storage
  • Datalogging and download capability on TSG205 via USB port to support clinical record management requirements


Warranty: 5 years on the solid state refrigeration system and 2 years parts and labor

Specifications

Specifications

2°C to 8°C
1.83 cu. ft.
120 V 60 Hz
NEMA 5-15 P
CRS Powder Coated
No
23 in.
584.2 mm
20.5 in.
22.1 in.
Solid
1
Countertop
TUV-SUD tested to cULus 61010-1 standard
51.8 L
120 V
USB Data Port
Right
309.88 mm
18.4 in.
Automatic
521 mm
561 mm
Built-in temperature and alarm event logging, USB export
3
SDS
Documents
Product Certifications
Promotions

Promotions