missing translation for 'onlineSavingsMsg'
Learn More

Tryptophol, 97%

CAS: 526-55-6 | C10H11NO | 161.20 g/mol

$64.40 - $64.40

Chemical Identifiers

CAS 526-55-6
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.20
MDL Number MFCD00005659
InChI Key MBBOMCVGYCRMEA-UHFFFAOYSA-N
Synonym tryptophol, 3-2-hydroxyethyl indole, indole-3-ethanol, 1h-indole-3-ethanol, 2-1h-indol-3-yl ethanol, 3-indoleethanol, indoleethanol, 3-indolylethanol, indole ethanol, 2-3-indolyl ethanol
PubChem CID 10685
ChEBI CHEBI:17890
SMILES OCCC1=CNC2=CC=CC=C12
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC140600010
SDS View Documents
Thermo Scientific Chemicals
140600010
1 g Glass bottle
Each for $64.40
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

526-55-6
161.20
MBBOMCVGYCRMEA-UHFFFAOYSA-N
10685
OCCC1=CNC2=CC=CC=C12
C10H11NO
MFCD00005659
tryptophol, 3-2-hydroxyethyl indole, indole-3-ethanol, 1h-indole-3-ethanol, 2-1h-indol-3-yl ethanol, 3-indoleethanol, indoleethanol, 3-indolylethanol, indole ethanol, 2-3-indolyl ethanol
CHEBI:17890

Specifications

526-55-6
White to Brown
255°C
96% min. (GC)
C10H11NO
1 g
15, 9978
Solubility in water: 10g/L (20°C). Other solubilities: soluble in glacial acetic acid, soluble in methanol, ethanol, acetone, chloroform, and ethyl acetate
OCCC1=CNC2=CC=CC=C12
161.20
CHEBI:17890
97%
Tryptophol, 97%
57.0°C to 60.0°C
174.0°C (2.7 mbar)
Authentic
Glass bottle
MFCD00005659
21, I, 218
tryptophol, 3-2-hydroxyethyl indole, indole-3-ethanol, 1h-indole-3-ethanol, 2-1h-indol-3-yl ethanol, 3-indoleethanol, indoleethanol, 3-indolylethanol, indole ethanol, 2-3-indolyl ethanol
MBBOMCVGYCRMEA-UHFFFAOYSA-N
2-(1H-indol-3-yl)ethanol
10685
161.2
Crystalline Mass, Crystals, Flakes or Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 208-393-2

RTECSNumber : KL3685000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only