missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tryptophol, 97%

Manufacturer:  Alfa Aesar™ L0255506

 View more versions of this product

Catalog No. AAL0255506



Description

Description

Applications
It is employed as a reactant for preparation of inhibitors of the C-terminal domain of RNA polymerase II and their antitumor activities, anti-HIV-1 agents, inhibitors of Protein-Protein Interactions, partial agonists of the serotonin 5-HT1A receptor, growth hormone secretagogues, vascular endothelial growth factor (VEGF) inhibitors, A2B adenosine receptor ligands, potential detoxification inhibitors of the crucifer phytoalexin brassinin, inhibitors of interleukin and dual binding site acetyl cholinesterase inhibitors.

Solubility
Solubility in methanol gives very faint turbidity. Soluble in water at 25°C 2749 mg/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

Tryptophol
174°C (2mmHg)
5g
C10H11NO
125553
1h-indole-3-ethanol
C1=CC=C2C(=C1)C(=CN2)CCO
161.204
CHEBI:17890
97%
255°C (491°F)
56°C to 59°C
526-55-6
MFCD00005659
14,9798
MBBOMCVGYCRMEA-UHFFFAOYSA-N
2-(1H-indol-3-yl)ethanol
10685
161.2

Chemical Identifiers

526-55-6
161.204
MBBOMCVGYCRMEA-UHFFFAOYSA-N
10685
2-(1H-indol-3-yl)ethanol
C10H11NO
MFCD00005659
1h-indole-3-ethanol
CHEBI:17890
C1=CC=C2C(=C1)C(=CN2)CCO
Safety and Handling

Safety and Handling

RTECSNumber : KL3685000

TSCA : TSCA

Documents