missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Truxene 98.0+%, TCI America™
$49.78 - $418.06
Chemical Identifiers
CAS | 548-35-6 |
---|---|
Molecular Formula | C27H18 |
Molecular Weight (g/mol) | 342.441 |
MDL Number | MFCD00021180 |
InChI Key | YGPLLMPPZRUGTJ-UHFFFAOYSA-N |
PubChem CID | 68355 |
SMILES | C1C2=CC=CC=C2C3=C1C4=C(CC5=CC=CC=C54)C6=C3CC7=CC=CC=C76 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
T2752100MG
|
TCI America
T2752100MG |
100 mg |
Each for $49.78
|
|
|||||
T27521G
|
TCI America
T27521G |
1 g |
Each for $418.06
|
|
|||||
Chemical Identifiers
548-35-6 | |
342.441 | |
YGPLLMPPZRUGTJ-UHFFFAOYSA-N | |
C1C2=CC=CC=C2C3=C1C4=C(CC5=CC=CC=C54)C6=C3CC7=CC=CC=C76 |
C27H18 | |
MFCD00021180 | |
68355 |
Specifications
548-35-6 | |
Yellow | |
MFCD00021180 | |
truxene, truxen, .alpha.-truxene, 5h-tribenzo a,f,k trindene, 10,15-dihydro, benzene, tribenzylene, 10,15-dihydro-5h-tribenzo a,f,k triindene, 10,15-dihydro-5h-diindeno 1,2-a:1',2'-c fluorene, 5h-tribenzo a,f,k trindene,10,15-dihydro, tribenzylenebenzene | |
C1C2=CC=CC=C2C3=C1C4=C(CC5=CC=CC=C54)C6=C3CC7=CC=CC=C76 | |
68355 | |
≥98.0% (HPLC) | |
Truxene |
370°C | |
C27H18 | |
100 mg | |
YGPLLMPPZRUGTJ-UHFFFAOYSA-N | |
342.441 | |
342.44 | |
Crystalline Powder |
Safety and Handling
TSCA : No