Please login to your online account to display your discounted pricing
Trundle Wheel
Measures long distances in the classroom or outdoors.
$30.75 - $65.45
Specifications
For Use With (Application) Designed for measuring a longer distance in the classroom or outdoors
Material Plastic
View More Specs
Products
Catalog Number Mfr. No. Includes Type Price Quantity    
S90871
fisher science education
4555
Counter, adjustable handle, tire with Rubber tread, loud non-wearing clicking device Super Deluxe Trundle Wheel Each for $65.45
S94313
fisher science education
45550
Tire with Rubber tread, loud non-wearing clicking device Trundle Wheel Each for $30.75
Description & Specifications
Specifications
For Use With (Application) Designed for measuring a longer distance in the classroom or outdoors
Material Plastic
  • Weather-resistant, all-plastic construction
  • Loud, non-wearing clicking device is activated in either direction with each full meter revolution
  • Comes apart in two pieces for storage and is easily reassembled
  • Counter measures up to 99,999m with adjustable handle
  • Tire is equipped with a rubber tread to provide accurate readings