Please login to your online account to display your discounted pricing

Thermo Scientific™ Tru1I (MseI)


Cut at T^TAA sites with Tru1I (MseI) restriction enzyme, which performs best at 65°C in R buffer (Isoschizomers: MseI, Tru9I).

Manufacturer: thermo scientific™  ER0981

 View more versions of this product

Catalog No. FERER0981

  • / Each
 

Description & Specifications

Specifications

Product Name Tru1I (MseI)
Product Name Tru1I (MseI)
Quantity 300U
Units 300 U
Concentration 10U/µL
Quantity 300U
Item Description 10U/uL, 300U

Lambda DNA digested with TrulI (MseI), 1.4% agarose, 195 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

Highlights

  • Superior quality -stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities