Human TRPM8, Synthetic Peptide, Invitrogen™
Synthetic Peptide
Manufacturer:  Invitrogen GST00017P
Catalog No. PIGST00017P
Description
Description
TRPM8 Synthetic Peptide, GST00017P, from Invitrogen.
TRPM8 is a receptor-activated non-selective cation channel involved in detection of sensations such as coolness, by being activated by cold temperature below 25 degrees Celsius. TRPM8 is activated by icilin, eucalyptol, menthol, cold and modulation of intracellular pH and involved in menthol sensation. TRPM8 is permeable for monovalent cations sodium, potassium, and cesium and divalent cation calcium. TRPM8 has an expression pattern within normal human tissues that is mostly restricted to prostate epithelial cells. TRPM8 mRNA expression is at moderate levels in normal prostate tissue and appears to be elevated in prostate cancer. Notably, TRPM8 mRNA was also expressed in a number of nonprostatic primary tumors of breast, colon, lung, and skin origin, whereas transcripts encoding TRPM8 were hardly detected or not detected in the corresponding normal human tissues. GST00017P is the immunizing peptide for the antibody to TRPM8 (OSR00126W and OST00028W).GST00017P may be used as a control in various immunology applications.GST00017P maintain refrigerated at 2-8°C for up to 12 months. After reconstitution keep at -20°C. Avoid freeze and thaw cycles.Reconstitute with PBS or distilled water.
Specifications
Specifications
Synthetic Peptide
79054
Human
Control
peptide with no preservative
RUO
No preservative
Human TRPM8
Q7Z2W7
Peptide
Human
purified
50μg
Lyophilized
Y
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Transient receptor potential cation channel subfamily M member 8, transient receptor potential-p8, Trp-p8, long transient receptor potential channel 6
TRPM8
Control
Documents