missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Troxipide, TCI America™

Manufacturer:  TCI America T29095G

 View more versions of this product

Catalog No. T29095G



Specifications

Specifications

Troxipide
180°C
30751-05-4
MFCD00866876
YSIITVVESCNIPR-UHFFFAOYSA-N
3,4,5-trimethoxy-N-piperidin-3-ylbenzamide
5597
Crystal-Powder at 20°C
White
5g
C15H22N2O4
3,4,5-Trimethoxy-N-(3-piperidyl)benzamide
COC1=CC(=CC(=C1OC)OC)C(=O)NC2CCCNC2
294.351
294.35
≥98.0% (HPLC,T)

Chemical Identifiers

30751-05-4
294.351
YSIITVVESCNIPR-UHFFFAOYSA-N
5597
COC1=CC(=CC(=C1OC)OC)C(=O)NC2CCCNC2
C15H22N2O4
MFCD00866876
3,4,5-Trimethoxy-N-(3-piperidyl)benzamide
3,4,5-trimethoxy-N-piperidin-3-ylbenzamide
Safety and Handling

Safety and Handling

RTECSNumber : CV5792600

TSCA : No

Documents