Learn More
Tropylium tetrafluoroborate, 97%, Thermo Scientific Chemicals
$354.48 - $3604.88
Chemical Identifiers
CAS | 27081-10-3 |
---|---|
Molecular Formula | C7H7BF4 |
Molecular Weight (g/mol) | 177.94 |
MDL Number | MFCD00013402 |
InChI Key | SQVQHTIKOZVGLR-UHFFFAOYSA-N |
Synonym | tropylium tetrafluoroborate, cycloheptatrienyl tetrafluoroborate, cyclohepta-1,3,5-triene tetrafluoroborate, cycloheptatrienylium, tetrafluoroborate 1-, tropylium tetrafluoroborate 1-, cycloheptatrienylium, tetrafluoroborate 1-1:1, cyclohepta-2,4,6-trienylium tetrafluoroborat, tropylium tetrafluoroborate, purum t |
PubChem CID | 152926 |
IUPAC Name | cyclohepta-1,3,5-triene;tetrafluoroborate |
SMILES | F[B-](F)(F)F.[CH+]1C=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1404506
|
Thermo Scientific Chemicals
A1404506 |
5 g |
Each for $354.48
|
|
|||||
AAA1404514
|
Thermo Scientific Chemicals
A1404514 |
25 g |
Each for $1,176.05
|
|
|||||
AAA1404522
|
Thermo Scientific Chemicals
A1404522 |
100 g |
Each for $3,604.88
|
|
|||||
Description
Reactant or reagent involved in Retro-Buchner reactions for synthesis of arylcyclopropanes, α-Cyanation of imines, electrochemical redox transformations, oxidative C-H activation of aliphatic aldehydes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsReactant or reagent involved in Retro-Buchner reactions for synthesis of arylcyclopropanes, α-Cyanation of imines, electrochemical redox transformations, oxidative C-H activation of aliphatic aldehydes.
Solubility
Soluble in hot methanol.
Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agents, strong bases, water/ moisture.
Chemical Identifiers
27081-10-3 | |
177.94 | |
SQVQHTIKOZVGLR-UHFFFAOYSA-N | |
152926 | |
F[B-](F)(F)F.[CH+]1C=CC=CC=C1 |
C7H7BF4 | |
MFCD00013402 | |
tropylium tetrafluoroborate, cycloheptatrienyl tetrafluoroborate, cyclohepta-1,3,5-triene tetrafluoroborate, cycloheptatrienylium, tetrafluoroborate 1-, tropylium tetrafluoroborate 1-, cycloheptatrienylium, tetrafluoroborate 1-1:1, cyclohepta-2,4,6-trienylium tetrafluoroborat, tropylium tetrafluoroborate, purum t | |
cyclohepta-1,3,5-triene;tetrafluoroborate |
Specifications
27081-10-3 | |
C7H7BF4 | |
5 g | |
3574512 | |
tropylium tetrafluoroborate, cycloheptatrienyl tetrafluoroborate, cyclohepta-1,3,5-triene tetrafluoroborate, cycloheptatrienylium, tetrafluoroborate 1-, tropylium tetrafluoroborate 1-, cycloheptatrienylium, tetrafluoroborate 1-1:1, cyclohepta-2,4,6-trienylium tetrafluoroborat, tropylium tetrafluoroborate, purum t | |
SQVQHTIKOZVGLR-UHFFFAOYSA-N | |
cyclohepta-1,3,5-triene;tetrafluoroborate | |
152926 | |
97% |
∼240°C (decomposition) | |
MFCD00013402 | |
UN1759 | |
Moisture sensitive | |
Soluble in hot methanol. | |
F[B-](F)(F)F.[CH+]1C=CC=CC=C1 | |
177.94 | |
177.94 | |
Tropylium tetrafluoroborate |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H500
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 248-214-5
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only