missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tropylium tetrafluoroborate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1404506

 View more versions of this product

Catalog No. AAA1404506



Description

Description

Reactant or reagent involved in Retro-Buchner reactions for synthesis of arylcyclopropanes, α-Cyanation of imines, electrochemical redox transformations, oxidative C-H activation of aliphatic aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant or reagent involved in Retro-Buchner reactions for synthesis of arylcyclopropanes, α-Cyanation of imines, electrochemical redox transformations, oxidative C-H activation of aliphatic aldehydes.

Solubility
Soluble in hot methanol.

Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agents, strong bases, water/ moisture.
Specifications

Specifications

Tropylium tetrafluoroborate
∼240°C (decomposition)
MFCD00013402
UN1759
Moisture sensitive
Soluble in hot methanol.
F[B-](F)(F)F.[CH+]1C=CC=CC=C1
177.94
177.94
27081-10-3
C7H7BF4
5 g
3574512
tropylium tetrafluoroborate, cycloheptatrienyl tetrafluoroborate, cyclohepta-1,3,5-triene tetrafluoroborate, cycloheptatrienylium, tetrafluoroborate 1-, tropylium tetrafluoroborate 1-, cycloheptatrienylium, tetrafluoroborate 1-1:1, cyclohepta-2,4,6-trienylium tetrafluoroborat, tropylium tetrafluoroborate, purum t
SQVQHTIKOZVGLR-UHFFFAOYSA-N
cyclohepta-1,3,5-triene;tetrafluoroborate
152926
97%

Chemical Identifiers

27081-10-3
177.94
SQVQHTIKOZVGLR-UHFFFAOYSA-N
152926
F[B-](F)(F)F.[CH+]1C=CC=CC=C1
C7H7BF4
MFCD00013402
tropylium tetrafluoroborate, cycloheptatrienyl tetrafluoroborate, cyclohepta-1,3,5-triene tetrafluoroborate, cycloheptatrienylium, tetrafluoroborate 1-, tropylium tetrafluoroborate 1-, cycloheptatrienylium, tetrafluoroborate 1-1:1, cyclohepta-2,4,6-trienylium tetrafluoroborat, tropylium tetrafluoroborate, purum t
cyclohepta-1,3,5-triene;tetrafluoroborate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H500

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 248-214-5

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only