missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Tropomyosin Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Tropomyosin Ab-1, Mouse Monoclonal Antibody.

$246.55 - $1,242.49

Specifications

Antigen Tropomyosin Ab-1
Clone CH1
Applications Immunofluorescence,Immunohistochemistry (Paraffin),Immunofluorescence,Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
MS1256P Thermo Scientific™ Lab Vision™
MS1256P
Purified Ab with BSA and Azide 1mL Each for $1,156.12
MS1256P0 Thermo Scientific™ Lab Vision™
MS1256P0
Purified Ab with BSA and Azide 100μL Each for $246.55
MS1256P1 Thermo Scientific™ Lab Vision™
MS1256P1
Purified Ab with BSA and Azide 500μL Each for $718.45
MS1256P1ABX Thermo Scientific™ Lab Vision™
MS1256P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $814.20
MS1256PABX Thermo Scientific™ Lab Vision™
MS1256PABX
Ab without BSA/Azide (Frozen) 200μL Each for $1,242.49
MS1256R7 Thermo Scientific™ Lab Vision™
MS1256R7
Ready-to-use for IHC (UVLP) 7mL Each for $684.88
Specifications

Description

Provide accurate, reproducible results with the Thermo Scientific™ Tropomyosin Ab-1, Mouse Monoclonal Antibody.

Tropomyosin is a dimeric coiled-coil protein that binds along the length of actin filaments. It is associated with the thin filaments of muscle cells and the microfilaments of nonmuscle cells. Chicken embryo fibroblasts (CEF) contain five isoforms of tropomyosin (a, b, 1, 2, and 3), identified as such by their different apparent molecular masses after separation by SDS-PAGE, but similar biochemical properties, such as resistance to heat and organic solvents, the ability to bind to F-actin filaments, and the lack of proline and tryptophan.

Host Species: Mouse

Clone: CH1

Isotype: IgG1/κ

Species Reactivity: Human, Rat and Chicken. Others not known.

Epitope: Not determined

Immunogen: Tropomyosin protein purified from chicken embryo fibroblasts (CEF) and chicken leg, heart and gizzard

Positive Control: Skeletal or Cardiac muscle

Cellular Localization: Cytoplasmic Ab-1 reacts strongly with striated muscle forms of tropomyosin, including cardiac alpha-tropomyosin and skeletal alpha- and Beta-tropomyosins. ^

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not verified)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Tropomyosin Ab-1
Immunofluorescence,Immunohistochemistry (Paraffin),Immunofluorescence,Immunohistochemistry (Paraffin)
Unlabeled
Murine
Muscle Biology
Tropomyosin protein purified from chicken embryo fibroblasts (CEF) and chicken leg, heart, and gizzard
CH1
Monoclonal
Chicken,Human,Rat,Chicken,Human,Rat,Chicken,Human,Rat
RUO
Primary
IgG2a kappa