Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Tropomyosin Ab-1, Mouse Monoclonal Antibody


Provide accurate, reproducible results with the Thermo Scientific™ Tropomyosin Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1256P1ABX

 View more versions of this product

Catalog No. MS1256P1ABX

  • / Each
 

Description & Specifications

Specifications

Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Immunogen Tropomyosin protein purified from chicken embryo fibroblasts (CEF) and chicken leg, heart, and gizzard
Isotype IgG2a kappa
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Rat
Quantity 100µL
Regulatory Status RUO
Antigen Tropomyosin Ab-1
Host Species Mouse
Clone CH1
Format Concentrated
Formulation Ab without BSA/Azide (Frozen)
Type Primary
Classification Monoclonal
Concentration 1mg/mL
Conjugate Unlabeled
Research Discipline Muscle Biology
Item Description 100uL; 1mg/mL; Purified w/o BSA/ azide

Provide accurate, reproducible results with the Thermo Scientific™ Tropomyosin Ab-1, Mouse Monoclonal Antibody.

Tropomyosin is a dimeric coiled-coil protein that binds along the length of actin filaments. It is associated with the thin filaments of muscle cells and the microfilaments of nonmuscle cells. Chicken embryo fibroblasts (CEF) contain five isoforms of tropomyosin (a, b, 1, 2, and 3), identified as such by their different apparent molecular masses after separation by SDS-PAGE, but similar biochemical properties, such as resistance to heat and organic solvents, the ability to bind to F-actin filaments, and the lack of proline and tryptophan.

Host Species: Mouse

Clone: CH1

Isotype: IgG1/κ

Species Reactivity: Human, Rat and Chicken. Others not known.

Epitope: Not determined

Immunogen: Tropomyosin protein purified from chicken embryo fibroblasts (CEF) and chicken leg, heart and gizzard

Positive Control: Skeletal or Cardiac muscle

Cellular Localization: Cytoplasmic Ab-1 reacts strongly with striated muscle forms of tropomyosin, including cardiac alpha-tropomyosin and skeletal alpha- and Beta-tropomyosins. ^

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not verified)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO