Please login to your online account to display your discounted pricing

Alfa Aesar™ Tropolone, 98%

As a reagent for microdetermination of rhodium (III)

Chemical Identifiers

Chemical Name or Material Tropolone
CAS 533-75-5
Formula Weight 122.12g/mol
MDL Number MFCD00004158
Synonym 2-Hydroxy-2,4,6-cycloheptatrienone; 2-Hydroxy-2,4,6-cycloheptatrien-1-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1652602

 
alfa aesar™
16526-02
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1652604

 
alfa aesar™
16526-04
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 0.5g
Melting Point 49° to 50°C
Flash Point >110° (230°F)
Quantity 0.5g
Chemical Name or Material Tropolone
CAS 533-75-5
Physical Form Crystalline
Assay 98%
Formula Weight 122.12g/mol
MDL Number MFCD00004158
Synonym 2-Hydroxy-2,4,6-cycloheptatrienone; 2-Hydroxy-2,4,6-cycloheptatrien-1-one