Learn More
Tropine DL-tropate, 99%, Thermo Scientific Chemicals
Reduces brain levels of acetylcholine
$46.30 - $746.49
Chemical Identifiers
CAS | 51-55-8 |
---|---|
Molecular Formula | C17H23NO3 |
Synonym | DL-Hyoscyamine; Atropine |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6467714
|
Thermo Scientific Chemicals
J6467714 |
25 g |
Each for $746.49
|
|
|||||
AAJ6467703
|
Thermo Scientific Chemicals
J6467703 |
1 g |
Each for $46.30
|
|
|||||
Description
Tropine DL-tropate is used as a reduces brain levels of acetylcholine, Competitive nonselective antagonist at central and peripheral muscarinic acetylcholine receptors, used in opthalmology to dilate the pupil and in efforts of resuscitation in the treatment of bradycardia and cardiac arrest.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
51-55-8 | |
C17H23NO3 | |
DL-Hyoscyamine; Atropine |
Specifications
25 g | |
Reduces brain levels of acetylcholine | |
C17H23NO3 | |
DL-Hyoscyamine; Atropine | |
Signal Transduction Reagent | |
91260 | |
CN1C2CCC1CC(C2)OC(=O)C(CO)C1=CC=CC=C1 | |
289.38 |
White | |
51-55-8 | |
289.37g/mol | |
Light Sensitive | |
Powder | |
RKUNBYITZUJHSG-UHFFFAOYNA-N | |
8-methyl-8-azabicyclo[3.2.1]octan-3-yl 3-hydroxy-2-phenylpropanoate | |
99% |
Safety and Handling
EINECSNumber : 200-104-8
Recommended Storage : Ambient temperatures
RUO – Research Use Only