Learn More
Tropine, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 140561000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Tropine | |
1.0400g/mL | |
Authentic | |
Glass bottle | |
MFCD00005551 | |
21, 16 | |
15, 9960 | |
Solubility in water: 100g/L (20°C). Other solubilities: soluble in ether and chloroform, freely soluble in alcohol | |
CN1C2CCC1CC(C2)O | |
141.21 | |
141.21 |
120-29-6 | |
233.0°C | |
98% | |
C8H15NO | |
100g | |
1.04 | |
tropine, 1r,5r-8-methyl-8-azabicyclo 3.2.1 octan-3-ol, endo-8-methyl-8-azabicyclo 3.2.1 octan-3-ol, spectrum3_001840, 1r,5r-8-methyl-8-azabicyclo 3.2.1 octan-3beta-ol | |
CYHOMWAPJJPNMW-RNFRBKRXSA-N | |
(1R,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-ol | |
6101956 | |
98% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
Do not eat,drink or smoke when using this product.
Avoid breathing dust/fume/
GHS Signal Word: Warning
EINECSNumber : 204-384-2
RUO â Research Use Only