Please login to your online account to display your discounted pricing

Alfa Aesar™ Tropaeolin O

Chemical Identifiers

Chemical Name or Material Tropaeolin O
CAS 547-57-9
Formula Weight 316.27
MDL Number MFCD00007499
Merck Index 14,9775
Synonym Acid Orange 6; C.I. 14270
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3870804

  SDS
alfa aesar™
3870804
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3870809

  SDS
alfa aesar™
3870809
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 316.27
Chemical Name or Material Tropaeolin O
Quantity 2g
TSCA TSCA
Physical Form Powder
MDL Number MFCD00007499
CAS 547-57-9
Merck Index 14,9775
Synonym Acid Orange 6; C.I. 14270
Solubility Information Soluble in water and alcohol
Color Orange to brown
RTECS Number DB6145180