Please login to your online account to display your discounted pricing

Tromethamine, Multi-Compendial, U.S.P., J.T.Baker™

$151.52 - $981.52

Chemical Identifiers

Chemical Name or Material Tromethamine
CAS 77-86-1
Purity Grade Multi-Compendial/USP
Molecular Formula NH2C(CH2OH)3
Formula Weight 121.14
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004524

 
avantor performance
410205
Poly Bottle 2.5kg Each for $981.52

02004525

 
avantor performance
410201
Poly Bottle 500g Each for $151.52
Description & Specifications

Specifications

Purity Grade Multi-Compendial/USP
Formula Weight 121.14
Chemical Name or Material Tromethamine
Molecular Formula NH2C(CH2OH)3
CAS 77-86-1
Packaging Poly Bottle
Quantity 2.5kg