missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trolamine, NF, 99-107.4%, Spectrum™ Chemical
TR143, 102-71-6, C6H15NO3
Supplier: Spectrum Chemical Mfg Cor TR1434LTGLCS4
Description
Spectrum™ Chemical Trolamine, NF is used as an emulsifying excipient in hand and body lotions, shaving creams, soaps, and shampoos. All Spectrum Chemical NF products, including all NF Trolamine products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 102-71-6 | |
| 0.0005 | |
| C6H15NO3 | |
| 4 x 4 L | |
| OCCN(CCO)CCO | |
| 149.19 |
| 1 | |
| Amber Glass Bottle | |
| 1.481-1.486 | |
| GSEJCLTVZPLZKY-UHFFFAOYSA-N | |
| 2-[bis(2-hydroxyethyl)amino]ethan-1-ol | |
| 99 to 107.4% |
Chemical Identifiers
| 102-71-6 | |
| 149.19 | |
| 2-[bis(2-hydroxyethyl)amino]ethan-1-ol |
| C6H15NO3 | |
| GSEJCLTVZPLZKY-UHFFFAOYSA-N | |
| OCCN(CCO)CCO |