missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Troemner™ Weight Cases

Used to protect electronic balance calibration weights

$129.70 - $220.43

Specifications

Quantity Each
View More Specs

Products 2
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
36-101-2752
View Documents
Troemner™
8C1000
Replacement Case for 10 g to 500 g Individual Weights
Each for $129.70
 
02-214-160
View Documents
Troemner™
8C5000
Replacement Case for 1 kg to 5 kg Individual Weights
Each for $220.43
 
Description

Description

  • Can hold individual weights anywhere from 5kg to 10g

Specifications

Specifications

Each
SDS