Please login to your online account to display your discounted pricing

Troemner™ UltraClass Platinum Individual Analytical Precision Weights with Traceable Certificate


For calibrating high-precision analytical balances with readability as low as 0.0001mg

Manufacturer: troemner llc  7028UCPT

 View more versions of this product

Catalog No. 02-214-413

  • / Each
 

Description & Specifications

Specifications

Product Type Metric analytical sheet weight with one side turned up
Weight (Metric) 200mg
Classification UltraClass Platinum
Includes Polycarbonate case
Certifications/Compliance Traceable certificate, ISO9001
Material 304 Stainless Steel
Quantity Each
Density 7.95g/cm3 at 20°C
Mass 200mg
Shape Flat with one end turned up for easy handling