Troemner™ UltraClass Platinum Individual Analytical Precision Weights with NVLAP Certificate
For calibrating high-precision analytical balances with readability as low as 0.0001mg
$1,071.49 - $7,950.34
Specifications
Certifications/Compliance NVLAP, ISO9001
Classification UltraClass Platinum
Includes Polycarbonate case
Quantity Each
View More Specs
Products 28
Catalog Number Mfr. No. Product Type Mass Shape Price Quantity  
Catalog Number Mfr. No. Product Type Mass Shape Price Quantity  
02-214-372 View Documents Troemner™
7014UCPW
Metric analytical weight with handling knob 500g Cylindrical with handling knob Each for $2,431.74
02-214-384 View Documents Troemner™
7010UCPW
Metric analytical weight with handling knob 5kg Cylindrical with handling knob Each for $7,950.34
02-214-390 View Documents Troemner™
7027UCPW
Metric analytical sheet weight with one side turned up 300mg Flat with one end turned up for easy handling Each for $1,145.46
02-214-393 View Documents Troemner™
7015UCPW
Metric analytical weight with handling knob 300g Cylindrical with handling knob Each for $2,258.77
02-214-396 View Documents Troemner™
7031UCPW
Metric analytical sheet weight with one side turned up 30mg Flat with one end turned up for easy handling Each for $1,145.46
02-214-399 View Documents Troemner™
7019UCPW
Metric analytical weight with handling knob 30g Cylindrical with handling knob Each for $1,262.80
02-214-402 View Documents Troemner™
7035UCPW
Metric analytical sheet weight with one side turned up 3mg Flat with one end turned up for easy handling Each for $1,145.46
02-214-405 View Documents Troemner™
7011UCPW
Metric analytical weight with handling knob 3kg Cylindrical with handling knob Each for $7,304.91
02-214-408 View Documents Troemner™
7023UCPW
Metric analytical weight with handling knob 3g Cylindrical with handling knob Each for $1,262.80
02-214-414 View Documents Troemner™
7016UCPW
Metric analytical weight with handling knob 200g Cylindrical with handling knob Each for $2,154.17
02-214-420 View Documents Troemner™
7020UCPW
Metric analytical weight with handling knob 20g Cylindrical with handling knob Each for $1,181.69
02-214-426 View Documents Troemner™
7012UCPW
Metric analytical weight with handling knob 2kg Cylindrical with handling knob Each for $5,623.71
02-214-432 View Documents Troemner™
7029UCPW
Metric analytical sheet weight with one side turned up 100mg Flat with one end turned up for easy handling Each for $1,206.17
02-214-438 View Documents Troemner™
7033UCPW
Metric analytical sheet weight with one side turned up 10mg Flat with one end turned up for easy handling Each for $1,128.26
02-214-447 View Documents Troemner™
7013UCPW
Metric analytical weight with handling knob 1kg Cylindrical with handling knob Each for $4,373.91
02-214-450 View Documents Troemner™
7025UCPW
Metric analytical weight with handling knob 1g Cylindrical with handling knob Each for $1,244.31
02-214-369 View Documents Troemner™
7026UCPW
Metric analytical sheet weight with one side turned up 500mg Flat with one end turned up for easy handling Each for $1,071.49
02-214-375 View Documents Troemner™
7030UCPW
Metric analytical sheet weight with one side turned up 50mg Flat with one end turned up for easy handling Each for $1,339.34
02-214-378 View Documents Troemner™
7018UCPW
Metric analytical weight with handling knob 50g Cylindrical with handling knob Each for $1,578.51
02-214-381 View Documents Troemner™
7034UCPW
Metric analytical sheet weight with one side turned up 5mg Flat with one end turned up for easy handling Each for $1,071.49
02-214-387 View Documents Troemner™
7022UCPW
Metric analytical weight with handling knob 5g Cylindrical with handling knob Each for $1,181.69
02-214-411 View Documents Troemner™
7028UCPW
Metric analytical sheet weight with one side turned up 200mg Flat with one end turned up for easy handling Each for $1,071.49
02-214-417 View Documents Troemner™
7032UCPW
Metric analytical sheet weight with one side turned up 20mg Flat with one end turned up for easy handling Each for $1,339.34
02-214-423 View Documents Troemner™
7036UCPW
Metric analytical sheet weight with one side turned up 2mg Flat with one end turned up for easy handling Each for $1,339.34
02-214-429 View Documents Troemner™
7024UCPW
Metric analytical weight with handling knob 2g Cylindrical with handling knob Each for $1,244.31
02-214-435 View Documents Troemner™
7017UCPW
Metric analytical weight with handling knob 100g Cylindrical with handling knob Each for $1,431.29
02-214-441 View Documents Troemner™
7021UCPW
Metric analytical weight with handling knob 10g Cylindrical with handling knob Each for $1,244.31
02-214-444 View Documents Troemner™
7037UCPW
Metric analytical sheet weight with one side turned up 1mg Flat with one end turned up for easy handling Each for $1,544.66
Description
Description
Made of Troemner Alloy 8, a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage. Exclusive material has a density of 8.0 at 25°C (controlled to ±0.1%), is essentially nonmagnetic in the annealed condition, and will not transform to magnetic under any combination of temperature and cold work down to -195.5°C (-320°F).
  • Weight tolerance is equal to ANSI/ASTM E617 Class 000 and Class E0
  • This class is used as a reference standard for calibrating other reference standards and weights where the stability of the environment and careful handling are assured
  • Appropriate for calibrating high-precision analytical balances with a readability as low as 0.0001mg
  • Weights come with a Positive Calibration Tolerance Guarantee
Note: Fractional weights 5 to 500mg are made of stainless steel (except 30mg, which are aluminum). Weights 3mg and below are made of aluminum.
Certifications
With NVLAP™ weight calibration certificate.
Specifications
Specifications
NVLAP, ISO9001
UltraClass Platinum
Polycarbonate case
Each