Troemner™ UltraClass Platinum Individual Analytical Precision Weights with NVLAP Certificate
For calibrating high-precision analytical balances with readability as low as 0.0001mg
Manufacturer:  Troemner™ 7033UCPW
 View more versions of this product
Catalog No. 02-214-438
Description
Description
Certifications
With NVLAP™ weight calibration certificate.
Specifications
Specifications
Metric analytical sheet weight with one side turned up
UltraClass Platinum
10mg
10mg
Polycarbonate case
NVLAP, ISO9001
Stainless Steel
7.95g/cm3 at 20°C
Flat with one end turned up for easy handling
Each
Documents