Please login to your online account to display your discounted pricing

Troemner™ UltraClass Platinum Individual Analytical Precision Weights with NVLAP Certificate


For calibrating high-precision analytical balances with readability as low as 0.0001mg

Manufacturer: troemner llc  7013UCPW

 View more versions of this product

Catalog No. 02-214-447

  • / Each
 

Description & Specifications

Specifications

Classification UltraClass Platinum
Includes Polycarbonate case
Quantity Each
Certifications/Compliance NVLAP, ISO9001
Shape Cylindrical with handling knob
Product Type Metric analytical weight with handling knob
Material Alloy 8 Stainless Steel
Density 8.03g/cm3 at 20°C
Mass 1kg
Weight (Metric) 1kg