Please login to your online account to display your discounted pricing

Troemner™ UltraClass Platinum Individual Analytical Precision Weights


For calibrating high-precision analytical balances with readability as low as 0.0001mg

Manufacturer: troemner llc  7036UCP

 View more versions of this product

Catalog No. 02-214-424

  • / Each
 

Description & Specifications

Specifications

Density 2.7g/cm3 at 20°C
Material 3003-H14 Aluminum
Classification UltraClass Platinum
Includes Polycarbonate case
Quantity Each
Mass 2mg
Certifications/Compliance Statement of accuracy, ISO9001
Weight (Metric) 2mg
Shape Flat with one end turned up for easy handling
Product Type Metric analytical sheet weight with one side turned up