Troemner™ UltraClass Platinum Electronic Balance Calibration Weights with Traceable Certificate
Constructed of special Troemner Alloy 8 stainless steel
Manufacturer:  Troemner™ 8042UCPT
 View more versions of this product
Catalog No. 02-218-113
Description
Description
Certifications
With Traceable Certificate.
Specifications
Specifications
Two-Piece Electronic Balance Calibration Weight
UltraClass Platinum
150g
150g
Polypropylene case
Traceable certificate, ISO9001
Stainless Steel
7.95g/cm3 at 20°C
Straight cylinder
Each
Documents