Troemner™ UltraClass Platinum Electronic Balance Calibration Weights with Traceable Certificate
Constructed of special Troemner Alloy 8 stainless steel
$407.09 - $8,891.71
Specifications
Certifications/Compliance Traceable certificate, ISO9001
Classification UltraClass Platinum
Product Type Two-Piece Electronic Balance Calibration Weight
Quantity Each
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Mass Shape Price Quantity  
Catalog Number Mfr. No. Includes Mass Shape Price Quantity  
02-218-108 Troemner Llc
8052UCPT
Polypropylene case 50g Straight cylinder Each for $407.09
02-218-110 Troemner Llc
8044UCPT
Polypropylene case 100g Straight cylinder Each for $435.17
02-218-119 Troemner Llc
8036UCPT
Polypropylene case 300g Straight cylinder Each for $668.46
02-218-122 Troemner Llc
8034UCPT
Polypropylene case 400g Straight cylinder Each for $745.60
02-218-124 Troemner Llc
8032UCPT
Polypropylene case 500g Straight cylinder Each for $737.20
02-218-127 Troemner Llc
8024UCPT
Polypropylene case 2kg Cylindrical with groove handle Each for $1,041.89
02-218-129 Troemner Llc
8016UCPT
Polypropylene case 5kg Cylindrical with groove handle Each for $2,062.46
02-218-133 Troemner Llc
8010UCPT
Without case 10kg Cylindrical with recessed grip handle Each for $4,257.03
02-218-102 Troemner Llc
8054UCPT
Polypropylene case 30g Straight cylinder Each for $407.09
02-218-105 Troemner Llc
8053UCPT
Polypropylene case 40g Straight cylinder Each for $419.91
02-218-113 Troemner Llc
8042UCPT
Polypropylene case 150g Straight cylinder Each for $595.60
02-218-116 Troemner Llc
8040UCPT
Polypropylene case 160g Straight cylinder Each for $595.60
02-218-131 Troemner Llc
8014UCPT
Polypropylene case 5kg Cylindrical with recessed grip handle Each for $3,196.57
02-218-136 Troemner Llc
8006UCPT
Without case 20kg Cylindrical with recessed grip handle Each for $6,440.31
02-218-139 Troemner Llc
8002UCPT
Without case 25kg Cylindrical with recessed grip handle Each for $7,854.34
02-218-142 Troemner Llc
8000UCPT
Without case 30kg Cylindrical with recessed grip handle Each for $8,891.71
Specifications
Description
UltraClass Platinum Series Weights combine high precision with the advantage of two-piece construction, avoiding costly replacement issues associated with one-piece weights. Consistent, automated robotic measurement means that Troemner UltraClass Series weights are the most precise two-piece weights with tolerances that are equal to ANSI/ASTM E617 Class 000 (UltraClass Platinum), Class 00 (UltraClass Gold) and Class 0 (UltraClass).
Two-piece weights with adjusting cavities
  • Made from stainless steel chosen for its uniformity density, high resistance to wear and corrosion, and low magnetic properties
  • UltraClass™ weights are highly polished to a mirror-like finish
Precision-Machined Stainless-Steel in Three Styles
  • Masses from 30 to 500g: straight cylinders
  • 1 to 5kg: cylindrical with indentation near top for easy gripping
  • 5 to 30kg: sleek cylinders with recessed grip handles, flat on top, easily stacked on center
Protective Cases
  • Individual weights 10g through 5kg supplied in high-quality polycarbonate case
  • Soft absorbent cotton glove is included to protect weights during use
  • Optional cases available for grip-handle weights
Certifications
With Traceable Certificate.
Description & Specifications
Specifications
Traceable certificate, ISO9001
UltraClass Platinum
Two-Piece Electronic Balance Calibration Weight
Each