Troemner™ UltraClass Platinum Electronic Balance Calibration Weights with NVLAP Certificate
Constructed of special Troemner Alloy 8 stainless steel
$574.97 - $8,081.74
Specifications
Certifications/Compliance NVLAP, ISO9001
Classification UltraClass Platinum
Product Type Two-Piece Electronic Balance Calibration Weight
Quantity Each
View More Specs
Products 10
Catalog Number Mfr. No. Includes Mass Shape Price Quantity  
Catalog Number Mfr. No. Includes Mass Shape Price Quantity  
02-218-101 View Documents Henry Troemner Inc
8054UCPW
Polypropylene case 30g Straight cylinder Each for $574.97
02-218-104 View Documents Henry Troemner Inc
8053UCPW
Polypropylene case 40g Straight cylinder Each for $605.09
02-218-107 View Documents Henry Troemner Inc
8052UCPW
Polypropylene case 50g Straight cylinder Each for $605.09
02-218-112 View Documents Henry Troemner Inc
8042UCPW
Polypropylene case 150g Straight cylinder Each for $940.80
02-218-115 View Documents Henry Troemner Inc
8040UCPW
Polypropylene case 160g Straight cylinder Each for $940.80
02-218-121 View Documents Henry Troemner Inc
8034UCPW
Polypropylene case 400g Straight cylinder Each for $940.80
02-218-130 View Documents Henry Troemner Inc
8014UCPW
Polypropylene case 5kg Cylindrical with recessed grip handle Each for $3,336.31
02-218-135 View Documents Henry Troemner Inc
8006UCPW
Without case 20kg Cylindrical with recessed grip handle Each for $6,718.20
02-218-138 View Documents Henry Troemner Inc
8002UCPW
Without case 25kg Cylindrical with recessed grip handle Each for $8,081.74
02-218-141 View Documents Henry Troemner Inc
8000UCPW
Without case 30kg Cylindrical with recessed grip handle N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
UltraClass Platinum Series Weights combine high precision with the advantage of two-piece construction, avoiding costly replacement issues associated with one-piece weights. Consistent, automated robotic measurement means that Troemner UltraClass Series weights are the most precise two-piece weights with tolerances that are equal to ANSI/ASTM E617 Class 000 (UltraClass Platinum), Class 00 (UltraClass Gold) and Class 0 (UltraClass).
Two-piece weights with adjusting cavities
  • Made from stainless steel chosen for its uniformity density, high resistance to wear and corrosion, and low magnetic properties
  • UltraClass™ weights are highly polished to a mirror-like finish
Precision-Machined Stainless-Steel in Three Styles
  • Masses from 30 to 500g: straight cylinders
  • 1 to 5kg: cylindrical with indentation near top for easy gripping
  • 5 to 30kg: sleek cylinders with recessed grip handles, flat on top, easily stacked on center
Protective Cases
  • Individual weights 10g through 5kg supplied in high-quality polycarbonate case
  • Soft absorbent cotton glove is included to protect weights during use
  • Optional cases available for grip-handle weights
Certifications
With NIST™/NVLAP™ Weight Calibration Certificate
Specifications
Specifications
NVLAP, ISO9001
UltraClass Platinum
Two-Piece Electronic Balance Calibration Weight
Each