Please login to your online account to display your discounted pricing
Troemner™ UltraClass Platinum Electronic Balance Calibration Weights
Constructed of special Troemner Alloy 8 stainless steel
$278.80 - $8,826.86
Specifications
Certifications/Compliance Statement of accuracy, ISO9001
Classification UltraClass Platinum
Product Type Two piece electronic balance calibration weight
Quantity Each
View More Specs
Products
Catalog Number Mfr. No. Includes Mass Shape Price Quantity    
02-218-106
troemner llc
8052UCP
Polypropylene case 50g Straight cylinder Each for $293.23
02-218-109
troemner llc
8044UCP
Polypropylene case 100g Straight cylinder Each for $357.23
02-218-114
troemner llc
8040UCP
Polypropylene case 160g Straight cylinder Each for $446.80
02-218-117
troemner llc
8038UCP
Polypropylene case 200g Straight cylinder Each for $429.74
02-218-120
troemner llc
8034UCP
Polypropylene case 400g Straight cylinder Each for $579.54
02-218-123
troemner llc
8032UCP
Polypropylene case 500g Straight cylinder Each for $617.63
02-218-125
troemner llc
8028UCP
Polypropylene case 1kg Cylindrical with groove handle Each for $735.23
02-218-132
troemner llc
8010UCP
Without case 10kg Cylindrical with recessed grip handle Each for $4,190.37
02-218-134
troemner llc
8006UCP
Without case 20kg Cylindrical with recessed grip handle Each for $6,223.40
02-218-100
troemner llc
8054UCP
Polypropylene case 30g Straight cylinder Each for $278.80
02-218-103
troemner llc
8053UCP
Polypropylene case 40g Straight cylinder Each for $300.23
02-218-111
troemner llc
8042UCP
Polypropylene case 150g Straight cylinder Each for $450.34
02-218-118
troemner llc
8036UCP
Polypropylene case 300g Straight cylinder Each for $497.54
02-218-126
troemner llc
8024UCP
Polypropylene case 2kg Cylindrical with groove handle Each for $862.09
02-218-128
troemner llc
8016UCP
Polypropylene case 5kg Cylindrical with groove handle Each for $1,947.23
02-218-137
troemner llc
8002UCP
Without case 25kg Cylindrical with recessed grip handle Each for $7,475.80
02-218-140
troemner llc
8000UCP
Without case 30kg Cylindrical with recessed grip handle Each for $8,826.86
Description & Specifications
Specifications
Certifications/Compliance Statement of accuracy, ISO9001
Classification UltraClass Platinum
Product Type Two piece electronic balance calibration weight
Quantity Each
UltraClass Platinum Series Weights combine high precision with the advantage of two-piece construction, avoiding costly replacement issues associated with one-piece weights. Consistent, automated robotic measurement means that Troemner UltraClass Series weights are the most precise two-piece weights with tolerances that are equal to ANSI/ASTM E617 Class 000 (UltraClass Platinum), Class 00 (UltraClass Gold) and Class 0 (UltraClass).
Two-piece weights with adjusting cavities
  • Made from stainless steel chosen for its uniformity density, high resistance to wear and corrosion, and low magnetic properties
  • UltraClass™ weights are highly polished to a mirror-like finish
Precision-Machined Stainless-Steel in Three Styles
  • Masses from 30 to 500g: straight cylinders
  • 1 to 5kg: cylindrical with indentation near top for easy gripping
  • 5 to 30kg: sleek cylinders with recessed grip handles, flat on top, easily stacked on center
Protective Cases
  • Individual weights 10g through 5kg supplied in high-quality polycarbonate case
  • Soft absorbent cotton glove is included to protect weights during use
  • Optional cases available for grip-handle weights