Troemner™ UltraClass Platinum Electronic Balance Calibration Weights
Constructed of special Troemner Alloy 8 stainless steel
Manufacturer:  Troemner™ 8016UCP
 View more versions of this product
Catalog No. 02-218-128
Specifications
Specifications
Two-Piece Electronic Balance Calibration Weight
UltraClass Platinum
5kg
5kg
Polypropylene case
Statement of accuracy, ISO9001
Stainless Steel
7.85g/cm3 at 20°C
Cylindrical with groove handle
Each
Documents