Troemner™ UltraClass Platinum Calibration Weight Sets with Traceable Certificate
For calibrating high-precision analytical balances with readability as low as 0.0001mg
Manufacturer:  Troemner™ 7217UCPT
 View more versions of this product
Catalog No. 02-214-454
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Certifications
With Traceable certificate.
Specifications
Specifications
Large metric analytical weight set
UltraClass Platinum
1mg to 1kg
Stainless Steel cylindrical weight: 1kg, 500g, 2 x 200g, 100g, 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, Stainless Steel sheet weight: 500mg, 2 x 200mg, 100mg, 50mg, 2 x 20mg, 10mg, 5mg, Aluminium sheet weight: 2 x 2mg, 1mg, Polycarbonate case in Wood case
Traceable certificate, ISO9001
Stainless Steel, Aluminum
1mg to 1kg
25/Set
Documents