Troemner™ UltraClass Platinum Calibration Weight Sets with Traceable Certificate
For calibrating high-precision analytical balances with readability as low as 0.0001mg
$5,285.77 - $9,620.89
Specifications
Certifications/Compliance Traceable certificate, ISO9001
Classification UltraClass Platinum
View More Specs
Products 25
Catalog Number Mfr. No. Material Weight (Metric) Mass Quantity Price Quantity  
Catalog Number Mfr. No. Material Weight (Metric) Mass Quantity Price Quantity  
02-214-454 View Documents Troemner™
7217UCPT
Stainless Steel, Aluminum 1mg to 1kg 1mg to 1kg 25/Set N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
02-214-457 View Documents Troemner™
7218UCPT
Stainless Steel 1g to 500g 500g-1g 12/Set N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
02-214-460 View Documents Troemner™
7219UCPT
Stainless Steel, Aluminum 1mg to 500g 1mg to 500g 24/Set N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
02-214-463 View Documents Troemner™
7220UCPT
Stainless Steel 1g to 200g 2-200g-1g 11/Set Each for $8,910.66
02-214-466 View Documents Troemner™
7221UCPT
Stainless Steel, Aluminum 1mg to 200g 1mg to (2)200g 23/Set N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
02-214-469 View Documents Troemner™
7222UCPT
Stainless Steel 1g to 200g 200g-1g 10/Set Each for $8,045.54
02-214-472 View Documents Troemner™
7223UCPT
Stainless Steel, Aluminum 1mg to 200g 1mg to 200g 22/Set N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
02-214-475 View Documents Troemner™
7224UCPT
Stainless Steel 1g to 100g 1 to 100g 9/Set Each for $6,006.63
02-214-478 View Documents Troemner™
7225UCPT
Stainless Steel 100mg to 100g 100g-100mg 13/Set Each for $8,751.86
02-214-481 View Documents Troemner™
7226UCPT
Stainless Steel 10mg to 100g 10mg to 100g 17/Set Each for $9,620.89
02-214-484 View Documents Troemner™
7227UCPT
Stainless Steel 5mg to 100g 100g-5mg 18/Set N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
02-214-487 View Documents Troemner™
7228UCPT
Stainless Steel, Aluminum 1mg to 100g 1mg to 100g 21/Set N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
02-214-490 View Documents Troemner™
7229UCPT
Stainless Steel 1g to 50g 1 to 50g 8/Set Each for $5,716.37
02-214-493 View Documents Troemner™
7230UCPT
Stainless Steel 100mg to 50g 50g-100mg 12/Set Each for $7,543.37
02-214-496 View Documents Troemner™
7231UCPT
Stainless Steel 10mg to 50g 10mg to 50g 16/Set N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
02-214-499 View Documents Troemner™
7232UCPT
Stainless Steel 5mg to 50g 50g-5mg 17/Set Each for $9,219.91
02-214-502 View Documents Troemner™
7233UCPT
Stainless Steel, Aluminum 1mg to 50g 50g-1mg 20/Set N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
02-214-508 View Documents Troemner™
7235UCPT
Stainless Steel 100mg to 20g 20g-100mg 10/Set Each for $6,681.03
02-214-505 View Documents Troemner™
7234UCPT
Stainless Steel 1g to 20g 20g-1g 6/Set Each for $5,319.89
02-214-511 View Documents Troemner™
7236UCPT
Stainless Steel 10mg to 20g 20g-10mg 14/Set Each for $8,194.29
02-214-514 View Documents Troemner™
7237UCPT
Stainless Steel 5mg to 20g 20g-5mg 15/Set Each for $7,515.40
02-214-517 View Documents Troemner™
7238UCPT
Stainless Steel, Aluminum 1mg to 20g 20g-1mg 18/Set Each for $8,751.86
02-214-520 View Documents Troemner™
7239UCPT
Stainless Steel 10mg to 500mg 500mg-10mg 8/Set Each for $5,285.77
02-214-523 View Documents Troemner™
7240UCPT
Stainless Steel, Aluminum 1mg to 500g 1 to 500mg 12/Set Each for $5,809.20
02-100-156 View Documents Troemner™
7214UCPT
Stainless Steel 1g to 2kg 1g to 2kg 14/Set N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Made of Troemner Alloy 8, a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage. Exclusive material has a density of 8.0 at 25°C (controlled to ±0.1%), is essentially nonmagnetic in the annealed condition, and will not transform to magnetic under any combination of temperature and cold work down to -195.5°C (-320°F).
  • Weight tolerance is equal to ANSI/ASTM E617 Class 000 and Class E0
  • This class is used as a reference standard for calibrating other reference standards and weights where the stability of the environment and careful handling are assured
  • Appropriate for calibrating high-precision analytical balances with a readability as low as 0.0001mg
  • Weights come with a Positive Calibration Tolerance Guarantee
Construction:
  • Fractional weights 5 to 500mg are made of stainless steel (except 30mg, which are aluminum)
  • Weights 3mg and below are made of aluminum
Note: Weight sets where the largest weight is 500g and below are supplied in a high-quality polycarbonate case. A wood case is provided for sets where the largest weight is 1kg and above.
Certifications
With Traceable certificate.
Specifications
Specifications
Traceable certificate, ISO9001
UltraClass Platinum